Learn More
eMolecules​ 34880-70-1 | 1-Methoxy-1-trimethylsilyloxypropene | Oakwood Chemical | MFCD00671554 | 160.288 | C7H16O2Si | 0.000 | CO\C(O[Si](C)(C)C)=C/C | 1g | 537724922
Catalog No. 5000158494 Shop All eMolecules Inc. Products
Encompass_Preferred
1-Methoxy-1-trimethylsilyloxypropene | Oakwood Chemical | 34880-70-1 | MFCD00671554 | 160.288 | C7H16O2Si | 0.000 | CO\C(O[Si](C)(C)C)=C/C | 1g | 537724922
Catalog No. 50-001-58494 Supplier eMolecules​ Supplier No. 537724922
1-Methoxy-1-trimethylsilyloxypropene | Oakwood Chemical | 34880-70-1 | MFCD00671554 | 160.288 | C7H16O2Si | 0.000 | CO\C(O[Si](C)(C)C)=C/C | 1g | 537724922