Learn More
World Precision Instrument 34g Flexible Quartz Tubing for filling
SDP
Catalog No. 502521059
Encompass
This tubing is used with a NanoFil syringe, most often for use in filling glass capillary pipettes.
Catalog No. 50-252-1059 Supplier World Precision Instrument Supplier No. NFQ345
This tubing is used with a NanoFil syringe, most often for use in filling glass capillary pipettes.