Learn More
Teknova 34mg/mL Chloramphenicol. 20mL, Sterile. 3/1170/II(L) - 1L limited
SDP
Catalog No. 502388148
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-8148 Supplier Teknova Supplier No. C0322
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.