Learn More
Teknova 34mg/mL Chloramphenicol Solution in Ethanol. 100mL, Sterile. 3/1170/II(L) - 1L limited
SDP
Catalog No. 502388053
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-8053 Supplier Teknova Supplier No. C0314
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.