Learn More
Thermo Scientific Dionex ASE 150/350 Cellulose Thimbles | | Pack of 25
Catalog No. NC0843207 Shop All Thermo Scientific Products
Encompass
- High-purity cellulose thimbles hold solid and semisolid samples in the extraction system, prevent plugging, and provide reliable and reproducible results. - Made from alpha cellulose, these thimbles have excellent mechanical strength. - The thimbles' dimensions accurately match Dionex ASE 350/150 System Extraction Cells to ensure an optimum fit during accelerated solvent extraction. - Simplifies cell cleaning when extracting sticky samples or samples that melt at high temperatures. - Enables direct weighing of samples and avoids sample loss during transfer. - Prevents fine particulate samples from clogging the cell.
Catalog No. NC0843207 Supplier Thermo Scientific Supplier No. 088349
- High-purity cellulose thimbles hold solid and semisolid samples in the extraction system, prevent plugging, and provide reliable and reproducible results. - Made from alpha cellulose, these thimbles have excellent mechanical strength. - The thimbles' dimensions accurately match Dionex ASE 350/150 System Extraction Cells to ensure an optimum fit during accelerated solvent extraction. - Simplifies cell cleaning when extracting sticky samples or samples that melt at high temperatures. - Enables direct weighing of samples and avoids sample loss during transfer. - Prevents fine particulate samples from clogging the cell.