Learn More
Environmental Sampling Supply Inc 34OZ OBLONG WIDE MOUTH 36/CS
Catalog No. NC1232415
Encompass_Preferred
34OZ HDPE WIDEMOUTH OBLONG CAPSURE/44MM POLY CLOSURE, EPA CERTIFIED
Catalog No. NC1232415 Supplier Environmental Sampling Supply Inc Supplier No. 10001902QC
34OZ HDPE WIDEMOUTH OBLONG CAPSURE/44MM POLY CLOSURE, EPA CERTIFIED