Learn More
Ace Glass, Inc. 35/20 14.5mm ID 25cm chromatography column with 250mLreservoir, stopcock, and porosity B disc.
SDP
Catalog No. 50972582
Encompass_Preferred
14.5mm I.D., 25cm chromatography column with a 35/20 joint and a 250L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom, and a porosity B (70-100 micron) fritted disc sealed in.
Catalog No. 50-972-582 Supplier Ace Glass, Inc. Supplier No. 590636
14.5mm I.D., 25cm chromatography column with a 35/20 joint and a 250L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom, and a porosity B (70-100 micron) fritted disc sealed in.