Learn More
Ace Glass, Inc. 35/20 25mm ID 30cm chromatography column with 250L reservoir and stopcock.
SDP
Catalog No. 50972579
Encompass_Preferred
25mm I.D., 30cm chromatography column with a 35/20 joint and a 250L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom.
Catalog No. 50-972-579 Supplier Ace Glass, Inc. Supplier No. 590614
25mm I.D., 30cm chromatography column with a 35/20 joint and a 250L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom.