Learn More
Chemglass Life Sciences 35/20 Ball Joint, Clamp Size: 35, Tubing OD: 21.70-22.30mm
SDP
Catalog No. CG12009
Encompass_Preferred
35/20 Ball Joint, Clamp Size: 35, Tubing OD: 21.70-22.30mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12009 Supplier Chemglass Life Sciences Supplier No. CG12009
35/20 Ball Joint, Clamp Size: 35, Tubing OD: 21.70-22.30mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.