Learn More
Chemglass Life Sciences 35/20 O-Ring Ball Joint, Clamp Size: 35, O-Ring Size: 120, Tubing OD: 21.70-22.30mm
SDP
Catalog No. CG12208
Encompass_Preferred
35/20 O-Ring Ball Joint, Clamp Size: 35, O-Ring Size: 120,Tubing OD: 21.70-22.30mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12208 Supplier Chemglass Life Sciences Supplier No. CG12208
35/20 O-Ring Ball Joint, Clamp Size: 35, O-Ring Size: 120,Tubing OD: 21.70-22.30mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.