Learn More
Chemglass Life Sciences Joint, 35/20, Polished Socket, Clamp Size 35, 21.70-22.30mm Tubing
SDP
Catalog No. CG121P05
Encompass_Preferred
Joint, 35/20, Polished Socket, Clamp Size 35, 21.70-22.30mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
Catalog No. CG121P05 Supplier Chemglass Life Sciences Supplier No. CG121P05
Joint, 35/20, Polished Socket, Clamp Size 35, 21.70-22.30mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.