Learn More
Ace Glass, Inc. 35/20 SOCKET JOINT (GLASSBLOWER)
SDP
Catalog No. 50975274
Encompass_Preferred
Full length, standard wall 35/20 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-274 Supplier Ace Glass, Inc. Supplier No. 765548
Full length, standard wall 35/20 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.