Learn More
Chemglass Life Sciences Joint, 35/20, Socket, Clamp Size 35, 21.70-22.30mm Tubing
SDP
Catalog No. CG12109
Encompass_Preferred
Joint, 35/20, Socket, Clamp Size 35, 21.70-22.30mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12109 Supplier Chemglass Life Sciences Supplier No. CG12109
Joint, 35/20, Socket, Clamp Size 35, 21.70-22.30mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.