Learn More
Ace Glass, Inc. 35/25 10mm straight adapter, glass only
SDP
Supplier:  Ace Glass, Inc. 806633
Encompass_Preferred
35/25 10mm straight adapter, glass portion only, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-979-641