Learn More
Ace Glass, Inc. Complete 35/25 10mm bearing, straight adapter style with hose connection port
SDP
Catalog No. 50975971
Encompass_Preferred
35/25 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-975-971 Supplier Ace Glass, Inc. Supplier No. 8066140
35/25 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.