Learn More
Ace Glass, Inc. Adapter, 105°, 35/25 socket top and ball bottom
SDP
Catalog No. 50971697
Encompass_Preferred
Angle adapter with 105 degree 35/25 socket outer top joint and 35/25 ball inner bottom joint.
Catalog No. 50-971-697 Supplier Ace Glass, Inc. Supplier No. 507535
Angle adapter with 105 degree 35/25 socket outer top joint and 35/25 ball inner bottom joint.