Learn More
Ace Glass, Inc. 35/25 ADAPTER GAS INLET 35/25 ADAPTER GAS INLET
SDP
Catalog No. 50978319
Encompass_Preferred
Side tube with hose connection. Stem below joint is 175mm. Hose Conn. size D. Available in 24/40 and 29/42 standard joints or 35/25 spherical joints.
Catalog No. 50-978-319 Supplier Ace Glass, Inc. Supplier No. 519035
Side tube with hose connection. Stem below joint is 175mm. Hose Conn. size D. Available in 24/40 and 29/42 standard joints or 35/25 spherical joints.