Learn More
Ace Glass, Inc. 35/25 BALL JOINT (GLASSBLOWER)
SDP
Catalog No. 50975275
Encompass_Preferred
Full length, standard wall 35/25 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-275 Supplier Ace Glass, Inc. Supplier No. 765552
Full length, standard wall 35/25 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.