Learn More
Ace Glass, Inc. 35/25 ball joint, stainless steel, unthreaded
SDP
Catalog No. 50975299
Encompass
35/25, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-975-299 Supplier Ace Glass, Inc. Supplier No. 765850
35/25, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.