Learn More
Ace Glass, Inc. 35/25 MALE ADAPT VAC CONN 35/25 MALE ADAPT VAC CONN
SDP
Catalog No. 50975478
Encompass_Preferred
Vacuum hose connecting adapter with 35/25 ball joint, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-478 Supplier Ace Glass, Inc. Supplier No. 781870
Vacuum hose connecting adapter with 35/25 ball joint, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.