Learn More
Ace Glass, Inc. Adapter, 35/25 socket joint, 3/4" tube end
SDP
Catalog No. 50978331
Encompass_Preferred
35/25 socket joint adapter with 3/4 tube end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
Catalog No. 50-978-331 Supplier Ace Glass, Inc. Supplier No. 522120
35/25 socket joint adapter with 3/4 tube end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.