Learn More
Ace Glass, Inc. Adapter, 35/25 socket joint, #25 Ace-Thred end
SDP
Catalog No. 50971776
Encompass_Preferred
35/25 socket joint adapter with #25 Ace-Thred end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
Catalog No. 50-971-776 Supplier Ace Glass, Inc. Supplier No. 522128
35/25 socket joint adapter with #25 Ace-Thred end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.