Learn More
Chemglass Life Sciences 35/25 Socket Joint, 28.0mm OD Tube, Clamp Size = 35
SDP
Catalog No. 501531915
Encompass_Preferred
35/25 Socket Joint, 28.0mm OD Tube, Clamp Size = 35. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. 50-153-1915 Supplier Chemglass Life Sciences Supplier No. CGB12110
35/25 Socket Joint, 28.0mm OD Tube, Clamp Size = 35. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.