Learn More
Chemglass Life Sciences Joint, 35/25, Socket, Clamp Size 35, 28mm Tubing
SDP
Catalog No. CG12110
Encompass_Preferred
Joint, 35/25, Socket, Clamp Size 35, 28mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12110 Supplier Chemglass Life Sciences Supplier No. CG12110
Joint, 35/25, Socket, Clamp Size 35, 28mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.