Learn More
Ace Glass, Inc. 35/25 socket joint, stainless steel, 1" male NPT
SDP
Catalog No. 50975309
Encompass
35/25 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1 male NPT.
Catalog No. 50-975-309 Supplier Ace Glass, Inc. Supplier No. 7658252
35/25 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1 male NPT.