Learn More
Ace Glass, Inc. 35/25 socket joint, stainless steel, unthreaded
SDP
Catalog No. 50979421
Encompass
35/25, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-979-421 Supplier Ace Glass, Inc. Supplier No. 765852
35/25, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.