Learn More
MSC 35 3/4 X66 GRN 3M ELECTRICAL
Catalog No. 501113903
Encompass
This electrical tape features a durable PVC backing and a pressure-sensitive, rubber-resin adhesive. It is designed for electrical insulation and protection, with a dielectric strength of 1250 V/mil, and operates within a temperature range of 0 to 105 °C.
  • Offers excellent electrical and mechanical protection.
  • Flame-retardant and highly resistant to abrasion, moisture, alkalis, solvents, and many acids.
  • UL Listed, VDE, CSA Certified, and RoHS 2011/65/EU Compliant.
Catalog No. 50-111-3903 Supplier MSC Supplier No. 54041272
This electrical tape features a durable PVC backing and a pressure-sensitive, rubber-resin adhesive. It is designed for electrical insulation and protection, with a dielectric strength of 1250 V/mil, and operates within a temperature range of 0 to 105 °C.
  • Offers excellent electrical and mechanical protection.
  • Flame-retardant and highly resistant to abrasion, moisture, alkalis, solvents, and many acids.
  • UL Listed, VDE, CSA Certified, and RoHS 2011/65/EU Compliant.