Learn More
MSC Scotch 35 Electrical Tape, 3/4 in, 1 ea
Catalog No. 501113915
Encompass
Scotch® Vinyl Color Coding Electrical Tape 35 is a premium-grade vinyl electrical tape designed to provide durable insulation and fade-resistant color-coding in demanding electrical applications. This versatile solution handles applications up to 600 V and endures temperatures from 14 °F to 221 °F (−10 °C to 105 °C).
  • Provides primary electrical insulation for splices up to 600 V.
  • Resists abrasion, moisture, chemicals, and varying weather conditions.
  • Maintains color integrity under UV and heat exposure.
  • Performs within temperatures from 14 °F to 221 °F (−10 °C to 105 °C).
  • Flexible PVC backing designed to conform to irregular shapes.
  • Bonds instantly to cable jackets for a reliable seal.
  • UL 510 Listed, CSA Certified, VDE Mark, IEC, and RoHS Compliant.
  • Flame retardant and cold resistant.
Catalog No. 50-111-3915 Supplier MSC Supplier No. 54041512
Scotch® Vinyl Color Coding Electrical Tape 35 is a premium-grade vinyl electrical tape designed to provide durable insulation and fade-resistant color-coding in demanding electrical applications. This versatile solution handles applications up to 600 V and endures temperatures from 14 °F to 221 °F (−10 °C to 105 °C).
  • Provides primary electrical insulation for splices up to 600 V.
  • Resists abrasion, moisture, chemicals, and varying weather conditions.
  • Maintains color integrity under UV and heat exposure.
  • Performs within temperatures from 14 °F to 221 °F (−10 °C to 105 °C).
  • Flexible PVC backing designed to conform to irregular shapes.
  • Bonds instantly to cable jackets for a reliable seal.
  • UL 510 Listed, CSA Certified, VDE Mark, IEC, and RoHS Compliant.
  • Flame retardant and cold resistant.