Learn More
Medchemexpress LLC 35-azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontan-1-ol | 1821464-55-4 | MFCD13184946 | 99.8% | C24H49N3O12 | 10MG
SDP
Catalog No. 5000261535
Encompass_Preferred
Azide-PEG12-alcohol is a polyethylene glycol (PEG)-based linker bearing a terminal azide and hydroxyl group. It is intended for use in bioconjugation and PROTAC synthesis, enabling efficient attachment to alkyne-containing partners via click chemistry (CuAAC and SPAAC).
  • High purity (≈99.8%) for reliable coupling reactions.
  • Long PEG12 spacer increases solubility and flexibility in aqueous systems.
  • Terminal azide functionality enables CuAAC and SPAAC click reactions.
  • Available in small-scale quantities suitable for research applications.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-61535 Supplier Medchemexpress LLC Supplier No. HY14080310MG
Azide-PEG12-alcohol is a polyethylene glycol (PEG)-based linker bearing a terminal azide and hydroxyl group. It is intended for use in bioconjugation and PROTAC synthesis, enabling efficient attachment to alkyne-containing partners via click chemistry (CuAAC and SPAAC).
  • High purity (≈99.8%) for reliable coupling reactions.
  • Long PEG12 spacer increases solubility and flexibility in aqueous systems.
  • Terminal azide functionality enables CuAAC and SPAAC click reactions.
  • Available in small-scale quantities suitable for research applications.
  • Stable under recommended storage conditions for extended shelf life.