Learn More
Apexbio Technology LLC 3,5-DHBA 99-10-5 5g
SDP
Catalog No. 502240431
Encompass_Preferred
3 5-Dihydroxybenzoic acid (3 5-DHBA CAS 99-10-5) is a naturally occurring phenolic compound identified as a biomarker for dietary intake of certain foods such as nuts beer peanuts and soy products Investigations indicate that 3 5-DHBA participates in cellular oxidative stress regulation likely via antioxidant pathways that influence the modulation of free radical levels and inflammatory signaling processes Recent studies have expanded its use in characterizing metabolic pathways exploring gut microbiota interactions and developing biomarkers for oxidative damage supporting research on the links between dietary components and metabolic health
This item is not returnable. View return policy
Catalog No. 50-224-0431 Supplier Apexbio Technology LLC Supplier No. B76865000
3 5-Dihydroxybenzoic acid (3 5-DHBA CAS 99-10-5) is a naturally occurring phenolic compound identified as a biomarker for dietary intake of certain foods such as nuts beer peanuts and soy products Investigations indicate that 3 5-DHBA participates in cellular oxidative stress regulation likely via antioxidant pathways that influence the modulation of free radical levels and inflammatory signaling processes Recent studies have expanded its use in characterizing metabolic pathways exploring gut microbiota interactions and developing biomarkers for oxidative damage supporting research on the links between dietary components and metabolic health