Learn More
Uline 3M 35 Electrical Tape - 34 x 66, Red
Catalog No. NC1903523
Encompass
Color-coded Quickly identify electrical circuits and leads, Splice and insulate wires up to 600V, Handles temperatures from 14F to 220F, Resists UV rays abrasion moisture copper corrosion alkalis and acids, Flame retardant, Polyvinyl chloride backing with rubber-based adhesive, UL Listed CSA certified
Catalog No. NC1903523 Supplier Uline Supplier No. S13975R
Color-coded Quickly identify electrical circuits and leads, Splice and insulate wires up to 600V, Handles temperatures from 14F to 220F, Resists UV rays abrasion moisture copper corrosion alkalis and acids, Flame retardant, Polyvinyl chloride backing with rubber-based adhesive, UL Listed CSA certified