Learn More
Biodesign Inc of Ny CellMicroSieves, 35 micron pore size, 2 x 3 foot (61 x 91.5 cm) roll
Catalog No. NC0027457
Encompass
CellMicroSieves (TM) filters manufactured by BioDesign Inc of New York allow the separation of multiple cell types using a filter that will not bind cells or proteins. Eleven pore sizes are available in sheets (3 sheets of 12 by 12 inches per package), rolls (2 feet by 3 feet), and 47mm circles (10 per package) that are extremely useful for specific cell type separation, stem cell isolation, hybridoma production, collection of cells from media, and many other uses where a strong non-binding specific sized pore filter is needed.  Can be repeatedly steam sterilized retaining its pore size.  A pure nylon thread product with no extractable materials that is specifially made for biologial research.
Catalog No. NC0027457 Supplier Biodesign Inc of Ny Supplier No. N35R
CellMicroSieves (TM) filters manufactured by BioDesign Inc of New York allow the separation of multiple cell types using a filter that will not bind cells or proteins. Eleven pore sizes are available in sheets (3 sheets of 12 by 12 inches per package), rolls (2 feet by 3 feet), and 47mm circles (10 per package) that are extremely useful for specific cell type separation, stem cell isolation, hybridoma production, collection of cells from media, and many other uses where a strong non-binding specific sized pore filter is needed.  Can be repeatedly steam sterilized retaining its pore size.  A pure nylon thread product with no extractable materials that is specifially made for biologial research.