Learn More
LGC GENOMICS LLC 35 ML COLUMN CHUCK
Supplier:  LGC GENOMICS LLC AXM121136
Encompass
Column holder for 35 mL synthesis columns (100 - 300 µmol), MerMade 6 and 12 only. Use with column M35-1000, scale dependent on support loading.
Catalog No. NC2310470