Learn More
CEM Corporation 35 ML VIAL WITH SIL CAP 5/PK
SDP
Catalog No. NC0211590
Encompass
Includes (5) 35mL vials, (5) 35mL ActiVent caps. NOTE: Caps are NOT compatible with the Discover S Class System.
Catalog No. NC0211590 Supplier CEM Corporation Supplier No. 909245
Includes (5) 35mL vials, (5) 35mL ActiVent caps. NOTE: Caps are NOT compatible with the Discover S Class System.