Learn More
Electron Microscopy Sciences 35 mm Biopsy Foam Pad
SDP
Catalog No. 5033120
Encompass_Preferred
A cellular polyester blue foam that has uniform air pockets throughout the pad for optimal fluid flow. It prevents either small or fragmented tissue specimens from being lost. To use simple place small samples or specimen fragments between the two pads and place the specimen in a cassette or capsule.
Catalog No. 50-331-20 Supplier Electron Microscopy Sciences Supplier No. 6232504
A cellular polyester blue foam that has uniform air pockets throughout the pad for optimal fluid flow. It prevents either small or fragmented tissue specimens from being lost. To use simple place small samples or specimen fragments between the two pads and place the specimen in a cassette or capsule.