Learn More
Cellvis 35 MM DISH WITH 14 MM M100/CS
Catalog No. NC2230478
Encompass
35 mm Dish with 14 mm micro-well and #1.5 glass-like polymer coverslip, tissue culture treated for better cell attachment than cover glass, 100/cs
This item is not returnable. View return policy
Catalog No. NC2230478 Supplier Cellvis Supplier No. D35141.5P
35 mm Dish with 14 mm micro-well and #1.5 glass-like polymer coverslip, tissue culture treated for better cell attachment than cover glass, 100/cs