Learn More
Ace Glass, Inc. Rotameter tube, size 35, with stainless steel float: 4.8 to50 liter/min air flow, no rating water flow, with glass float: 2.5-28 liter/min air flow, no rating water flow
SDP
Catalog No. 50974815
Encompass
Catalog No. 50-974-815 Supplier Ace Glass, Inc. Supplier No. 748123