Learn More
Cole-Parmer Environmental Express HotBlock SC151 Digestion System, 35-Well, 100 mL; 120 VAC
Catalog No. NC1101080 Shop All Cole-Parmer Products
Encompass
This is an EPA-approved system that simplifies and improves metals prep and digestions. It features an integrated block heater and controller design. Blocks are PTFE coated for chemical and corrosion resistance. Users can set high and low temperature alarm limits. System includes polycarbonate transfer racks designed to set on the block.
Catalog No. NC1101080 Supplier Cole-Parmer Supplier No. 5320508
Add to Cart
Add to Cart
Request a Quote
This is an EPA-approved system that simplifies and improves metals prep and digestions. It features an integrated block heater and controller design. Blocks are PTFE coated for chemical and corrosion resistance. Users can set high and low temperature alarm limits. System includes polycarbonate transfer racks designed to set on the block.