Learn More
Chemglass Life Sciences Vacuum Trap, 35mm Body OD x 250mm Body Length, 16mm OD Side and Inner Tubes
SDP
Supplier:  Chemglass Life Sciences CG451003
Encompass_Preferred
Vacuum Trap, 35mm Body OD x 250mm Body Length, 16mm OD Side and Inner Tubes. One piece trap used in the removal of undesired vapors in the vacuum system by condensation. Vacuum Trap, 35mm Body OD x 250mm Body Length, 16mm OD Side and Inner Tubes.
Catalog No. CG451003