Learn More
Accuflow Systems Altafluor 350 THV Flex - has the flexibility of traditional PVC with chemical resistance to FEP - 350 THV 1/8" X 1/4"- 25 FT
Catalog No. 501355124
Encompass_Preferred
Altafluor 350 THV FLEX (Tetrafluoroethylene-hexafluoropropylene. The preferred choice for UV light transmission. Good chemical resistance. Excellent bondability to other materials and has excellent ozone resistance, low permeation with superior weatherability.
Catalog No. 50-135-5124 Supplier Accuflow Systems Supplier No. 350025006225
Altafluor 350 THV FLEX (Tetrafluoroethylene-hexafluoropropylene. The preferred choice for UV light transmission. Good chemical resistance. Excellent bondability to other materials and has excellent ozone resistance, low permeation with superior weatherability.