Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 3500xL Dx Genetic Analyzer 24-Capillary Array, 50 cm

Catalog No. 4404688
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Array
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-046-88 1 Array
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-046-88 Supplier Applied Biosystems™ Supplier No. 4404688
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Ambient, 1 array with 24 capillaries

A radio frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.

These internally uncoated 50-cm capillaries are supplied in assemblies of 24 capillaries per array, with a built-in frame for easy installation. Radio frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The array can be used for 160 injections in Dx mode.

This product is intended for in vitro diagnostic use on the 3500xL Dx Genetic Analyzer and is not validated for use with other systems or applications. Refer to the 3500 Dx Genetic Analyzer User Guide for further details.

These internally uncoated 50cm capillaries are supplied in preassembled sets assemblies of 24 capillaries per array, with a built-in frame for easy installation.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Capillary Length 50 cm
Content And Storage Ambient, 1 array with 24 capillaries
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Product Type 3500xL Dx Genetic Analyzer
Quantity 1 Array
Shipping Condition Room Temperature
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.