Learn More
Sterlitech Corporation Buchner Funnel, Porcelain, Capacity 350ML, Paper Size 90 mm
SDP
Catalog No. NC1437774
Encompass_Preferred
BUCHNER FUNNEL, PORCELAIN, CAPACITY 350ML, PAPER SIZE 90 MM
Catalog No. NC1437774 Supplier Sterlitech Corporation Supplier No. 1510024
BUCHNER FUNNEL, PORCELAIN, CAPACITY 350ML, PAPER SIZE 90 MM