Learn More
Chemglass Life Sciences 350mL Filter Funnel, Buchner, 29/42 Lower Vacuum Assembly, Coarse Frit
SDP
Catalog No. CG140627
Encompass_Preferred
350mL Filter Funnel, Buchner, 29/42 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140627 Supplier Chemglass Life Sciences Supplier No. CG140627
Add to Cart
Add to Cart
350mL Filter Funnel, Buchner, 29/42 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.