Learn More
Chemglass Life Sciences 350mL Filter Funnel, Jacketed, Medium Frit
SDP
Catalog No. CG140315
Encompass_Preferred
350mL Filter Funnel, Jacketed, Medium Frit. Similar to CG-1402 but with a water jacket enclosing the funnel bowl and upper portion of the stem. Jacket permits heating or cooling of the contents of the funnel during filtration. Stem length listed is the length below the jacket. Jacketed filter funnels are available in sizes other than those listed. Please call for details.
Catalog No. CG140315 Supplier Chemglass Life Sciences Supplier No. CG140315
350mL Filter Funnel, Jacketed, Medium Frit. Similar to CG-1402 but with a water jacket enclosing the funnel bowl and upper portion of the stem. Jacket permits heating or cooling of the contents of the funnel during filtration. Stem length listed is the length below the jacket. Jacketed filter funnels are available in sizes other than those listed. Please call for details.