Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, 350mL Capacity, 70mm ID X 125mm Inside Depth, 86mm OD X 157mm Height
SDP
Catalog No. CG159401
Encompass_Preferred
Dewar Flask, Wide Mouth, Metal Housing, 350mL Capacity, 70mm ID X 125mm Inside Depth, 86mm OD X 157mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159401 Supplier Chemglass Life Sciences Supplier No. CG159401
Dewar Flask, Wide Mouth, Metal Housing, 350mL Capacity, 70mm ID X 125mm Inside Depth, 86mm OD X 157mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.