Learn More
Ace Glass, Inc. 350mL Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
SDP
Supplier:  Ace Glass, Inc. 718654
Encompass_Preferred
350mL Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
Catalog No. 50-974-284