Learn More
Sigma Aldrich Fine Chemicals Biosciences 355-Trimethylhexanal 97 fG1KG
Catalog No. 502903223
Encompass
3 5 5-Trimethylhexanal is an aliphatic aldehyde mainly used as an internal standard to quantify the volatile aldehydes in the root tissues of plants.
Catalog No. 50-290-3223 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3524031KGK
3 5 5-Trimethylhexanal is an aliphatic aldehyde mainly used as an internal standard to quantify the volatile aldehydes in the root tissues of plants.