Learn More
Sigma Aldrich Fine Chemicals Biosciences 355 Trimethylhexanal 97 fG4KG
Catalog No. 502914362
Encompass
3 5 5-Trimethylhexanal is an aliphatic aldehyde mainly used as an internal standard to quantify the volatile aldehydes in the root tissues of plants.
Catalog No. 50-291-4362 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3524034KGK
3 5 5-Trimethylhexanal is an aliphatic aldehyde mainly used as an internal standard to quantify the volatile aldehydes in the root tissues of plants.