Learn More
Ace Glass, Inc. Double tube vacuum manifold, 355mm long, (3) stopcock ports with 24/40 inner joints
SDP
Catalog No. 50976289
Encompass_Preferred
355mm long double tube vacuum manifold with four #15 o-ring joint ends. Manifold has three stopcock ports with 24/40 inner joints and 0-10MM Easy Action stopcocks. Distance between ports is 100MM which will allow for flasks up to 250mL.
Catalog No. 50-976-289 Supplier Ace Glass, Inc. Supplier No. 873918
355mm long double tube vacuum manifold with four #15 o-ring joint ends. Manifold has three stopcock ports with 24/40 inner joints and 0-10MM Easy Action stopcocks. Distance between ports is 100MM which will allow for flasks up to 250mL.