Learn More
Ace Glass, Inc. Double tube vacuum manifold, 355mm long, (3) stopcock ports with #15 O-ring joints
SDP
Catalog No. 50979836
Encompass_Preferred
355mm long double tube vacuum manifold with four #15 o-ring joint ends. Manifold has three stopcock ports with #15 o-ring joints and 0-10MM Easy Action stopcocks. Distance between ports is 100MM which will allow for flasks up to 250mL.
Catalog No. 50-979-836 Supplier Ace Glass, Inc. Supplier No. 873921
355mm long double tube vacuum manifold with four #15 o-ring joint ends. Manifold has three stopcock ports with #15 o-ring joints and 0-10MM Easy Action stopcocks. Distance between ports is 100MM which will allow for flasks up to 250mL.